Assay Details
| Gene Symbol: Mybbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016776.2 | NP_058056.2 | 9 - 10 | 1323 | 74 | |
| GenBank mRNA | AK083908.1 | - | 9 - 10 | 1349 | 74 | |
| AK143469.1 | - | 9 - 10 | 1347 | 74 | ||
| AK143487.1 | - | 9 - 10 | 1356 | 74 | ||
| AK149747.1 | - | 4 - 5 | 632 | 74 | ||
| AK154016.1 | - | 8 - 9 | 1446 | 74 | ||
| AK155074.1 | - | 9 - 10 | 1334 | 74 | ||
| BC015070.1 | - | 9 - 10 | 1324 | 74 | ||
| BC048858.1 | - | 9 - 10 | 1323 | 5363740 | 74 | |
| BC052889.1 | - | 9 - 10 | 1344 | 30057169 | 74 | |
| BC060167.1 | - | 9 - 10 | 1329 | 74 | ||
| U63648.1 | - | 9 - 10 | 1332 | 74 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mybbp1a
Genomic Map