Assay Details
| Gene Symbol: Camkk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018883.2 | NP_061371.2 | 1 - 2 | 118 | 73 | |
| GenBank mRNA | AF117384.1 | - | 1 - 2 | 73 | 73 | |
| AK144483.1 | - | 1 - 2 | 119 | 73 | ||
| BC017529.1 | - | 1 - 2 | 83 | 4924656 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF117384.1, BC017529.1, NM_018883.2 transcripts.
Set Membership: |
|

Camkk1
Genomic Map