Assay Details
| Gene Symbol: Camkk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018883.2 | NP_061371.2 | 3 - 4 | 572 | 140 | |
| GenBank mRNA | AF117384.1 | - | 3 - 4 | 527 | 140 | |
| AK144483.1 | - | 3 - 4 | 573 | 140 | ||
| BC017529.1 | - | 3 - 4 | 537 | 4924656 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Camkk1
Genomic Map