Assay Details
| Gene Symbol: Camkk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018883.2 | NP_061371.2 | 7 - 8 | 850 | 62 | |
| GenBank mRNA | AK144483.1 | - | 7 - 8 | 851 | 62 | |
| BC017529.1 | - | 7 - 8 | 815 | 4924656 | 62 | |
More Information
Set Membership: |
|

Camkk1
Genomic Map