Assay Details
| Gene Symbol: Camkk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018883.2 | NP_061371.2 | 8 - 9 | 872 | 73 | |
| GenBank mRNA | AK144483.1 | - | 8 - 9 | 873 | 73 | |
| BC017529.1 | - | 8 - 9 | 837 | 4924656 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Camkk1
Genomic Map