| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100018014.mm.1 |
NM_001252532.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018014.mm.1 |
NM_001252532.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018014.mm.1 |
NM_145438.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018014.mm.1 |
NM_145438.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018014.mm.1 |
TC1100001908.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100018008.mm.1 |
NM_001252532.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018008.mm.1 |
NM_001252532.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018008.mm.1 |
NM_145438.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018008.mm.1 |
NM_145438.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018008.mm.1 |
TC1100001908.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100018009.mm.1 |
NM_001252532.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018009.mm.1 |
NM_001252532.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018009.mm.1 |
NM_145438.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018009.mm.1 |
NM_145438.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018009.mm.1 |
TC1100001908.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100018016.mm.1 |
NM_001252532.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018016.mm.1 |
NM_001252532.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018016.mm.1 |
NM_145438.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018016.mm.1 |
NM_145438.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018016.mm.1 |
TC1100001908.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100018013.mm.1 |
NM_001252532.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018013.mm.1 |
NM_001252532.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018013.mm.1 |
NM_145438.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018013.mm.1 |
NM_145438.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018013.mm.1 |
TC1100001908.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100018011.mm.1 |
NM_001252532.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018011.mm.1 |
NM_001252532.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018011.mm.1 |
NM_145438.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018011.mm.1 |
NM_145438.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018011.mm.1 |
TC1100001908.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100018015.mm.1 |
NM_001252532.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018015.mm.1 |
NM_001252532.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018015.mm.1 |
NM_145438.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018015.mm.1 |
NM_145438.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018015.mm.1 |
TC1100001908.mm.1 |
MTA TC |