Assay Details
| Gene Symbol: Caskin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080643.2 | NP_542374.2 | 16 - 17 | 2153 | 67 | |
| GenBank mRNA | AF451978.1 | - | 15 - 16 | 1872 | 67 | |
| AK037133.1 | - | 3 - 4 | 278 | 67 | ||
| AK129297.1 | - | 16 - 17 | 2191 | 67 | ||
| BC053083.1 | - | 16 - 17 | 2153 | 5704642 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Caskin2
Genomic Map