Assay Details
| Gene Symbol: Caskin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080643.2 | NP_542374.2 | 17 - 18 | 2295 | 62 | |
| GenBank mRNA | AF451978.1 | - | 16 - 17 | 2014 | 62 | |
| AK037133.1 | - | 4 - 5 | 420 | 62 | ||
| AK129297.1 | - | 17 - 18 | 2333 | 62 | ||
| BC053083.1 | - | 17 - 18 | 2295 | 5704642 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Caskin2
Genomic Map