Assay Details
| Gene Symbol: Gltpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146020.1 | NP_666132.1 | 2 - 3 | 442 | 65 | |
| GenBank mRNA | AK050107.1 | - | 2 - 3 | 456 | 65 | |
| AK050212.1 | - | 2 - 3 | 456 | 65 | ||
| BC025515.1 | - | 2 - 3 | 441 | 5309987 | 65 | |
| BC030735.1 | - | 2 - 3 | 442 | 5126172 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901137 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901139 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901133 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901135 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901136 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901134 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278266 | AK050212.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278266 | BC025515.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278266 | BC030735.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278266 | NM_146020.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278266 | AK050107.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278266 | AK050212.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278266 | BC025515.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278266 | BC030735.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278266 | NM_146020.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278266 | AK050107.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6101 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148154 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278266 | AK050212.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278266 | BC025515.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278266 | BC030735.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278266 | NM_146020.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278266 | AK050107.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278266 | AK050212.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278266 | BC025515.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278266 | BC030735.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278266 | NM_146020.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278266 | AK050107.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901134 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148154 | - | - |
| Whole Mouse Genome G4122A | A_51_P278266 | AK050212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278266 | BC025515.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278266 | BC030735.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278266 | NM_146020.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278266 | AK050107.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278266 | AK050212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278266 | BC025515.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278266 | BC030735.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278266 | NM_146020.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278266 | AK050107.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106834 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148154 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09578614 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09578617 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901133 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901139 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901135 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901134 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901137 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901136 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09686138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09560754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09578614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901127 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10377850 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377849 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377848 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10377847 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100008415.mm.1 | NM_146020.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008415.mm.1 | NM_146020.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008415.mm.1 | TC1100000954.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100008416.mm.1 | NM_146020.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008416.mm.1 | NM_146020.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008416.mm.1 | TC1100000954.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 439974 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3940128 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3940128 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003940128 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2664842 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2664842 | - | - |
| mouse-6 v1.1 (Illumina) | scl41336.2.1_23-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3940128 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41336.2.1_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3940128 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2664841 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2874836 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2874836 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2664841 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl41336.2.1_23-S | - | - |

Gltpd2
Genomic Map