Assay Details
| Gene Symbol: Eftud2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109995.1 | NP_001103465.1 | 20 - 21 | 2134 | 74 | |
| NM_011431.3 | NP_035561.1 | 20 - 21 | 2131 | 74 | ||
| GenBank mRNA | AK052242.1 | - | 20 - 21 | 2089 | 74 | |
| AK150158.1 | - | 20 - 21 | 2102 | 74 | ||
| AK153473.1 | - | 20 - 21 | 2103 | 74 | ||
| AK154878.1 | - | 20 - 21 | 2113 | 74 | ||
| AK165624.1 | - | 20 - 21 | 2105 | 74 | ||
| AK172875.1 | - | 20 - 21 | 2085 | 74 | ||
| AK192949.1 | - | 1 - 2 | 44 | 74 | ||
| AK202628.1 | - | 1 - 2 | 44 | 74 | ||
| AK202816.1 | - | 1 - 2 | 32 | 74 | ||
| AK212456.1 | - | 1 - 2 | 44 | 74 | ||
| BC012636.1 | - | 7 - 8 | 846 | 74 | ||
| BC052674.1 | - | 20 - 21 | 2107 | 30061580 | 74 | |
| BC054778.1 | - | 20 - 21 | 2097 | 6810127 | 74 | |
| U97079.1 | - | 20 - 21 | 2073 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eftud2
Genomic Map