Assay Details
| Gene Symbol: Bcl2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009743.5 | NP_033873.3 | 1 - 2 | 127 | 76 | |
| GenBank mRNA | AK150028.1 | - | 1 - 2 | 69 | 76 | |
| BB856100.1 | - | 1 - 2 | 130 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_009743.5 transcripts.
Set Membership: |
|

Bcl2l1
Genomic Map