Assay Details
| Gene Symbol: Fam101b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029658.1 | NP_083934.1 | 1 - 2 | 356 | 67 | |
| GenBank mRNA | AA016881.1 | - | 1 - 2 | 245 | 67 | |
| AK005164.1 | - | 1 - 2 | 358 | 67 | ||
| AK171075.1 | - | 1 - 2 | 323 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam101b
Genomic Map