Assay Details
| Gene Symbol: Fam101b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029658.1 | NP_083934.1 | 2 - 3 | 468 | 62 | |
| GenBank mRNA | AA016881.1 | - | 2 - 3 | 357 | 62 | |
| AK005164.1 | - | 2 - 3 | 470 | 62 | ||
| AK171075.1 | - | 2 - 3 | 435 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275697 | AK005164.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275697 | AK033150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275697 | AK171075.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275697 | NM_029658.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275697 | AK005164.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275697 | AK033150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275697 | AK171075.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275697 | NM_029658.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P5891 | AK033150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P5891 | NM_029658.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P5891 | AK033150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P5891 | NM_029658.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807900 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 105131 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167179 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275697 | AK005164.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275697 | AK033150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275697 | AK171075.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275697 | NM_029658.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275697 | AK005164.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275697 | AK033150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275697 | AK171075.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275697 | NM_029658.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P5891 | AK033150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P5891 | NM_029658.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P5891 | AK033150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P5891 | NM_029658.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02366145 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167179 | - | - |
| Whole Mouse Genome G4122A | A_51_P275697 | AK005164.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275697 | AK033150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275697 | AK171075.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275697 | NM_029658.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275697 | AK005164.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275697 | AK033150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275697 | AK171075.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275697 | NM_029658.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P5891 | AK033150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P5891 | NM_029658.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P5891 | AK033150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P5891 | NM_029658.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09415534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09323160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09533305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09522398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09522394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09647246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09158517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09522387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09311998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09497933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09497934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09189505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09158516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09158511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09374668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27929491 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1429764_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429765_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456603_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10388488 | - | - |
| Mouse Genome 430 2.0 Array | 1429764_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456603_at | - | - |
| Mouse Genome 430 2.0 Array | 1429765_at | - | - |
| Mouse Genome 430 2.0 Array | 1429764_at | - | - |
| Mouse Genome 430 2.0 Array | 1456603_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429765_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76566_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76566_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100029765.mm.1 | NM_029658.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100029765.mm.1 | NM_029658.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100029765.mm.1 | TC1100003236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100029764.mm.1 | NM_029658.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100029764.mm.1 | NM_029658.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100029764.mm.1 | TC1100003236.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456603_at | - | - |
| Mouse Expression Set 430 | 1456603_PM_at | - | - |
| Mouse Expression Set 430 | 1429765_at | - | - |
| Mouse Expression Set 430 | 1429765_PM_at | - | - |
| Mouse Expression Set 430 | 1429764_at | - | - |
| Mouse Expression Set 430 | 1429764_PM_at | - | - |
| Murine Genome U74v2 Set | 96791_at | - | - |
| Murine Genome U74v2 Set | 96791_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456603_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456603_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456603_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456603_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429764_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429765_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429765_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429764_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2490729 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1217606 | - | - |

Fam101b
Genomic Map