Assay Details
| Gene Symbol: Olfr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146921.2 | NP_667132.1 | 2 - 3 | 288 | 92 | |
| GenBank mRNA | DR065653.1 | - | 2 - 3 | 288 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_146921.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr1
Genomic Map