Assay Details
| Gene Symbol: Srp68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146032.3 | NP_666144.3 | 2 - 3 | 355 | 62 | |
| GenBank mRNA | AK033010.1 | - | 2 - 3 | 274 | 62 | |
| AK212446.1 | - | 2 - 3 | 149 | 62 | ||
| AK213152.1 | - | 2 - 3 | 149 | 62 | ||
| BC070422.1 | - | 2 - 3 | 355 | 6403747 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srp68
Genomic Map