Assay Details
| Gene Symbol: Sphk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172475.1 | NP_001165946.1 | 1 - 2 | 172 | 75 | |
| GenBank mRNA | AF068748.1 | - | 1 - 2 | 172 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001172475.1 transcripts.
Set Membership: |
|

Sphk1
Genomic Map