Assay Details
| Gene Symbol: Qrich2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033267.2 | NP_001028439.1 | 6 - 7 | 1013 | 90 | |
| GenBank mRNA | AK131945.1 | - | 6 - 7 | 1016 | 90 | |
| BC150976.1 | - | 6 - 7 | 866 | 9087889 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Qrich2
Genomic Map