Assay Details
| Gene Symbol: Mgat5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172948.3 | NP_766536.2 | 5 - 6 | 1133 | 94 | |
| GenBank mRNA | AB116028.1 | - | 5 - 6 | 520 | 94 | |
| AB119127.1 | - | 5 - 6 | 599 | 94 | ||
| BC094604.1 | - | 5 - 6 | 1133 | 6822690 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mgat5b
Genomic Map