Assay Details
Gene Symbol: Cdk3-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_004853.1 | 2 - 3 | 191 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | TEN1 telomerase capping complex subunit | Ten1 | 2310004N24Rik | -1.1kb | Sense | Chr. 11 - 116198855 - 116215318 on NCBI Build GRCm38 |
Downstream | envoplakin | Evpl | 3.9kb | Anti-sense | Chr. 11 - 116220559 - 116238147 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
