Assay Details
Gene Symbol: Olfr411 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146709.2 | NP_666920.2 | 1 | 146 | 92 | |
GenBank mRNA | BC138439.1 | - | 1 | 146 | 8861459 | 92 |
BC138440.1 | - | 1 | 146 | 8861460 | 92 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
