Assay Details
| Gene Symbol: Smyd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102611.1 | NP_001096081.1 | 6 - 7 | 2037 | 78 | |
| GenBank mRNA | AK030380.1 | - | 6 - 7 | 1874 | 78 | |
| AK089959.1 | - | 6 - 7 | 1973 | 78 | ||
| AK154507.1 | - | 5 - 6 | 1723 | 78 | ||
| AK173306.1 | - | 4 - 5 | 1459 | 78 | ||
| BC049810.1 | - | 6 - 7 | 1888 | 78 | ||
| BC095952.1 | - | 6 - 7 | 1934 | 5037842 | 78 | |
| BC130220.1 | - | 3 - 4 | 1374 | 40141368 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smyd4
Genomic Map