Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100039146.mm.1 |
NM_026280.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039146.mm.1 |
NM_026280.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039146.mm.1 |
TC1100004126.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100039143.mm.1 |
NM_026280.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039143.mm.1 |
NM_026280.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039143.mm.1 |
TC1100004126.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100039142.mm.1 |
NM_026280.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039142.mm.1 |
NM_026280.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039142.mm.1 |
TC1100004126.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100039144.mm.1 |
NM_026280.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039144.mm.1 |
NM_026280.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039144.mm.1 |
TC1100004126.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100039147.mm.1 |
TC1100004126.mm.1 |
MTA TC |