Assay Details
| Gene Symbol: Enpp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030291.1 | NP_001025462.1 | 1 - 2 | 279 | 54 | |
| GenBank mRNA | AK144474.1 | - | 1 - 2 | 282 | 54 | |
| AK167678.1 | - | 1 - 2 | 282 | 54 | ||
| BC141245.1 | - | 1 - 2 | 364 | 9055857 | 54 | |
| BC171967.1 | - | 1 - 2 | 364 | 9054253 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Enpp7
Genomic Map