Assay Details
| Gene Symbol: Enpp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030291.1 | NP_001025462.1 | 3 - 4 | 1043 | 74 | |
| GenBank mRNA | AK144474.1 | - | 3 - 4 | 1046 | 74 | |
| AK167678.1 | - | 3 - 4 | 1046 | 74 | ||
| BC141245.1 | - | 3 - 4 | 1128 | 9055857 | 74 | |
| BC171967.1 | - | 3 - 4 | 1128 | 9054253 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28175185 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28175187 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28175188 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28175186 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28175184 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999653 | BC141245.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999653 | AK167678.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999653 | NM_001030291.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999653 | AK144474.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 449109 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554644 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999653 | BC141245.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999653 | AK167678.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999653 | NM_001030291.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999653 | AK144474.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28175104 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07512283 | - | - |
| Whole Mouse Genome G4122A | A_52_P316396 | BC141245.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P316396 | BC171967.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P316396 | AK167678.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P316396 | NM_001030291.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P316396 | AK144474.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09340577 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09668585 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09340574 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09668590 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09668583 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09340575 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09340576 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28175184 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28175185 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28175187 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28175188 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28175186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28175103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09157042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09177904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09108562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09014151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09356524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09706173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09108563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09595938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09157041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175077 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09411239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09635999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09668590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09701859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09096796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09340577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09293835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09595937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09177908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28175076 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10383053 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383052 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383054 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383051 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383049 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383048 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383050 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10383047 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100018948.mm.1 | NM_001030291.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018948.mm.1 | NM_001030291.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018948.mm.1 | TC1100001992.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100018946.mm.1 | TC1100001992.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100018947.mm.1 | TC1100001992.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100018950.mm.1 | TC1100001992.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100018945.mm.1 | NM_001030291.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018945.mm.1 | NM_001030291.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018945.mm.1 | TC1100001992.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100018943.mm.1 | NM_001030291.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018943.mm.1 | NM_001030291.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018943.mm.1 | TC1100001992.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 824616 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2534526 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3154252 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3075190 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3075190 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3154252 | - | - |

Enpp7
Genomic Map