Assay Details
| Gene Symbol: Bcas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164369.1 | NP_001157841.1 | 9 - 10 | 1447 | 88 | |
| NM_029815.2 | NP_084091.2 | 10 - 11 | 1585 | 88 | ||
| GenBank mRNA | AK008957.1 | - | 7 - 8 | 991 | 88 | |
| AY219233.1 | - | 10 - 11 | 1568 | 88 | ||
| BC113166.1 | - | 9 - 10 | 1427 | 40059981 | 88 | |
| BC113957.1 | - | 9 - 10 | 1427 | 40059979 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcas1
Genomic Map