Assay Details
| Gene Symbol: Bcas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164369.1 | NP_001157841.1 | 10 - 11 | 1508 | 62 | |
| NM_029815.2 | NP_084091.2 | 11 - 12 | 1646 | 62 | ||
| GenBank mRNA | AK008957.1 | - | 8 - 9 | 1052 | 62 | |
| AY219233.1 | - | 11 - 12 | 1629 | 62 | ||
| BC024553.1 | - | 1 - 2 | 59 | 62 | ||
| BC048647.1 | - | 2 - 3 | 198 | 62 | ||
| BC113166.1 | - | 10 - 11 | 1488 | 40059981 | 62 | |
| BC113957.1 | - | 10 - 11 | 1488 | 40059979 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC048647.1 transcripts.
Set Membership: |
|

Bcas1
Genomic Map