Assay Details
| Gene Symbol: Bcas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164369.1 | NP_001157841.1 | 1 - 2 | 92 | 90 | |
| NM_029815.2 | NP_084091.2 | 1 - 2 | 92 | 90 | ||
| GenBank mRNA | AK020254.1 | - | 1 - 2 | 94 | 90 | |
| AY219233.1 | - | 1 - 2 | 75 | 90 | ||
| BC113166.1 | - | 1 - 2 | 72 | 40059981 | 90 | |
| BC113957.1 | - | 1 - 2 | 72 | 40059979 | 90 | |
| BY720653.1 | - | 1 - 2 | 94 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY219233.1, BC113166.1, BC113957.1, NM_001164369.1, NM_029815.2 transcripts.
Set Membership: |
|

Bcas1
Genomic Map