Assay Details
| Gene Symbol: Bcas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164369.1 | NP_001157841.1 | 5 - 6 | 982 | 73 | |
| NM_029815.2 | NP_084091.2 | 5 - 6 | 982 | 73 | ||
| GenBank mRNA | AK008957.1 | - | 2 - 3 | 388 | 73 | |
| AK031825.1 | - | 2 - 3 | 388 | 73 | ||
| AY219233.1 | - | 5 - 6 | 965 | 73 | ||
| BC113166.1 | - | 5 - 6 | 962 | 40059981 | 73 | |
| BC113957.1 | - | 5 - 6 | 962 | 40059979 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcas1
Genomic Map