Assay Details
| Gene Symbol: Bcas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029815.2 | NP_084091.2 | 6 - 7 | 1032 | 91 | |
| GenBank mRNA | AK008957.1 | - | 3 - 4 | 438 | 91 | |
| AK031825.1 | - | 3 - 4 | 438 | 91 | ||
| AY219233.1 | - | 6 - 7 | 1015 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bcas1
Genomic Map