Assay Details
| Gene Symbol: Bcas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164369.1 | NP_001157841.1 | 8 - 9 | 1286 | 97 | |
| NM_029815.2 | NP_084091.2 | 9 - 10 | 1424 | 97 | ||
| GenBank mRNA | AK008957.1 | - | 6 - 7 | 830 | 97 | |
| AY219233.1 | - | 9 - 10 | 1407 | 97 | ||
| BC113166.1 | - | 8 - 9 | 1266 | 40059981 | 97 | |
| BC113957.1 | - | 8 - 9 | 1266 | 40059979 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcas1
Genomic Map