Assay Details
| Gene Symbol: Bcas1os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040610.1 | 7 - 8 | 905 | 106 | ||
| GenBank mRNA | AK006663.1 | - | 7 - 8 | 905 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | silica-induced gene 111 | Gm14270 |
OTTMUSG00000016146, SIG-111, Silg111 |
-93.1kb | Sense | Chr. 2 - 170284957 - 170286052 on NCBI Build GRCm38 |
| Downstream |
cytochrome P450, family 24, subfamily a, polypeptide 1 |
Cyp24a1 |
24-OHase, CP24, Cyp24 |
102.5kb | Anti-sense | Chr. 2 - 170482965 - 170497138 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Bcas1os2
Genomic Map