Assay Details
| Gene Symbol: 1700018P08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006097.1 | - | 1 - 2 | 28 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | avian musculoaponeurotic fibrosarcoma (v-maf) AS42 oncogene homolog | Maf |
2810401A20Rik, A230108G15Rik, AW047063, c-maf |
-650.2kb | Anti-sense | Chr. 8 - 115703253 - 115706894 on NCBI Build GRCm38 |
| Downstream | dynein light chain roadblock-type 2 | Dynlrb2 |
1700009A04Rik, DNLC2B, Dncl2b |
145.5kb | Sense | Chr. 8 - 116505015 - 116515915 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1700018P08Rik
Genomic Map