Assay Details
| Gene Symbol: Gfap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001131020.1 | NP_001124492.1 | 4 - 5 | 849 | 120 | |
| NM_010277.3 | NP_034407.2 | 4 - 5 | 849 | 120 | ||
| GenBank mRNA | AF332061.1 | - | 4 - 5 | 676 | 120 | |
| AF332062.1 | - | 4 - 5 | 676 | 120 | ||
| AK079577.1 | - | 4 - 5 | 783 | 120 | ||
| AK140151.1 | - | 4 - 5 | 781 | 120 | ||
| BC100728.1 | - | 4 - 5 | 728 | 120 | ||
| BC100737.1 | - | 4 - 5 | 721 | 120 | ||
| BC101968.1 | - | 4 - 5 | 728 | 120 | ||
| BC103571.1 | - | 4 - 5 | 722 | 120 | ||
| BC139357.1 | - | 4 - 5 | 849 | 8862379 | 120 | |
| BC139358.1 | - | 4 - 5 | 849 | 8862380 | 120 | |
| CX219900.1 | - | 2 - 3 | 193 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gfap
Genomic Map