Assay Details
| Gene Symbol: Mroh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039557.4 | NP_001034646.2 | 3 - 4 | 620 | 118 | |
| GenBank mRNA | AK029507.1 | - | 3 - 4 | 621 | 118 | |
| AK089157.1 | - | 3 - 4 | 621 | 118 | ||
| AK132763.1 | - | 3 - 4 | 377 | 118 | ||
| BC150937.1 | - | 3 - 4 | 344 | 9087848 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mroh8
Genomic Map