Assay Details
| Gene Symbol: Mroh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039557.4 | NP_001034646.2 | 5 - 6 | 834 | 67 | |
| GenBank mRNA | AK029507.1 | - | 5 - 6 | 835 | 67 | |
| AK132763.1 | - | 5 - 6 | 591 | 67 | ||
| BC150937.1 | - | 5 - 6 | 558 | 9087848 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mroh8
Genomic Map