Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000010232.mm.1 |
NR_045342.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010232.mm.1 |
NR_045342.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010232.mm.1 |
TC1000001370.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000010236.mm.1 |
NR_045342.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010236.mm.1 |
NR_045342.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010236.mm.1 |
TC1000001370.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000010233.mm.1 |
NR_045342.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010233.mm.1 |
NR_045342.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010233.mm.1 |
TC1000001370.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000010234.mm.1 |
NR_045342.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010234.mm.1 |
NR_045342.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010234.mm.1 |
TC1000001370.mm.1 |
MTA TC |