Assay Details
| Gene Symbol: Slc10a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282108.1 | NP_001269037.1 | 6 - 7 | 721 | 110 | |
| NM_029736.2 | NP_084012.1 | 6 - 7 | 721 | 110 | ||
| NR_104094.1 | 5 - 6 | 638 | 110 | |||
| GenBank mRNA | AK010834.1 | - | 6 - 7 | 671 | 110 | |
| AK040517.1 | - | 6 - 7 | 600 | 110 | ||
| AK079901.1 | - | 6 - 7 | 698 | 110 | ||
| AK152752.1 | - | 6 - 7 | 682 | 110 | ||
| AK152849.1 | - | 6 - 7 | 682 | 110 | ||
| AY825926.1 | - | 6 - 7 | 474 | 110 | ||
| AY825927.1 | - | 5 - 6 | 391 | 110 | ||
| BC116666.1 | - | 6 - 7 | 587 | 40107077 | 110 | |
| BC116717.1 | - | 6 - 7 | 587 | 40107078 | 110 | |
More Information
Set Membership: |
|

Slc10a7
Genomic Map