Assay Details
| Gene Symbol: Slc10a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009981.2 | NP_001009981.1 | 7 - 8 | 876 | 69 | |
| NM_001282109.1 | NP_001269038.1 | 7 - 8 | 884 | 69 | ||
| NM_029736.2 | NP_084012.1 | 8 - 9 | 960 | 69 | ||
| NR_104094.1 | 7 - 8 | 877 | 69 | |||
| GenBank mRNA | AK010834.1 | - | 8 - 9 | 910 | 69 | |
| AK040517.1 | - | 8 - 9 | 839 | 69 | ||
| AK051650.1 | - | 5 - 6 | 722 | 69 | ||
| AK152752.1 | - | 8 - 9 | 921 | 69 | ||
| AK152849.1 | - | 8 - 9 | 921 | 69 | ||
| AK160402.1 | - | 8 - 9 | 944 | 69 | ||
| AK165983.1 | - | 7 - 8 | 821 | 69 | ||
| AY825926.1 | - | 8 - 9 | 713 | 69 | ||
| AY825927.1 | - | 7 - 8 | 630 | 69 | ||
| AY825928.1 | - | 7 - 8 | 629 | 69 | ||
| BC116666.1 | - | 8 - 9 | 826 | 40107077 | 69 | |
| BC116717.1 | - | 8 - 9 | 826 | 40107078 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc10a7
Genomic Map