Assay Details
| Gene Symbol: Abcb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019552.2 | NP_062425.1 | 11 - 12 | 2150 | 75 | |
| GenBank mRNA | AF266284.1 | - | 11 - 12 | 1880 | 75 | |
| AK081782.1 | - | 11 - 12 | 1862 | 75 | ||
| BC046818.1 | - | 11 - 12 | 2106 | 5702696 | 75 | |
| BC053020.1 | - | 11 - 12 | 2106 | 5702696 | 75 | |
| BC054793.1 | - | 11 - 12 | 2150 | 6833785 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcb10
Genomic Map