Assay Details
| Gene Symbol: Abcb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019552.2 | NP_062425.1 | 1 - 2 | 717 | 100 | |
| GenBank mRNA | AF266284.1 | - | 1 - 2 | 447 | 100 | |
| AK081782.1 | - | 1 - 2 | 429 | 100 | ||
| BC046818.1 | - | 1 - 2 | 673 | 5702696 | 100 | |
| BC053020.1 | - | 1 - 2 | 673 | 5702696 | 100 | |
| BC054793.1 | - | 1 - 2 | 717 | 6833785 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcb10
Genomic Map