Assay Details
| Gene Symbol: Abcb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019552.2 | NP_062425.1 | 10 - 11 | 2116 | 84 | |
| GenBank mRNA | AF266284.1 | - | 10 - 11 | 1846 | 84 | |
| AK081782.1 | - | 10 - 11 | 1828 | 84 | ||
| BC046818.1 | - | 10 - 11 | 2072 | 5702696 | 84 | |
| BC053020.1 | - | 10 - 11 | 2072 | 5702696 | 84 | |
| BC054793.1 | - | 10 - 11 | 2116 | 6833785 | 84 | |
More Information
Set Membership: |
|

Abcb10
Genomic Map