Assay Details
| Gene Symbol: Itgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010578.2 | NP_034708.1 | 10 - 11 | 1572 | 134 | |
| GenBank mRNA | AK076526.1 | - | 6 - 7 | 869 | 134 | |
| AK088016.1 | - | 10 - 11 | 1473 | 134 | ||
| AK088729.1 | - | 10 - 11 | 1319 | 134 | ||
| AK135521.1 | - | 9 - 10 | 1248 | 134 | ||
| AK141253.1 | - | 5 - 6 | 499 | 134 | ||
| AK159689.1 | - | 10 - 11 | 1414 | 134 | ||
| AK167682.1 | - | 10 - 11 | 1470 | 134 | ||
| BC050906.1 | - | 10 - 11 | 1445 | 5721348 | 134 | |
| Y00769.1 | - | 10 - 11 | 1360 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itgb1
Genomic Map