Assay Details
| Gene Symbol: Itgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010578.2 | NP_034708.1 | 12 - 13 | 2018 | 103 | |
| GenBank mRNA | AK076526.1 | - | 8 - 9 | 1315 | 103 | |
| AK088016.1 | - | 12 - 13 | 1920 | 103 | ||
| AK088729.1 | - | 12 - 13 | 1765 | 103 | ||
| AK135521.1 | - | 11 - 12 | 1694 | 103 | ||
| AK141253.1 | - | 7 - 8 | 945 | 103 | ||
| AK159689.1 | - | 12 - 13 | 1860 | 103 | ||
| AK167682.1 | - | 12 - 13 | 1916 | 103 | ||
| BC050906.1 | - | 12 - 13 | 1891 | 5721348 | 103 | |
| BC119428.2 | - | 2 - 3 | 103 | |||
| X15202.1 | - | 11 - 12 | 1714 | 103 | ||
| Y00769.1 | - | 12 - 13 | 1806 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itgb1
Genomic Map