Assay Details
| Gene Symbol: Itgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010578.2 | NP_034708.1 | 13 - 14 | 2242 | 81 | |
| GenBank mRNA | AK088016.1 | - | 13 - 14 | 2144 | 81 | |
| AK088729.1 | - | 13 - 14 | 1989 | 81 | ||
| AK135521.1 | - | 12 - 13 | 1918 | 81 | ||
| AK141253.1 | - | 8 - 9 | 1169 | 81 | ||
| AK159689.1 | - | 13 - 14 | 2084 | 81 | ||
| AK167682.1 | - | 13 - 14 | 2140 | 81 | ||
| BC050906.1 | - | 13 - 14 | 2115 | 5721348 | 81 | |
| BC119428.2 | - | 1 - 2 | 81 | |||
| X15202.1 | - | 12 - 13 | 1938 | 81 | ||
| Y00769.1 | - | 13 - 14 | 2030 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itgb1
Genomic Map