Assay Details
| Gene Symbol: Itgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010578.2 | NP_034708.1 | 7 - 8 | 1239 | 86 | |
| GenBank mRNA | AK076526.1 | - | 3 - 4 | 536 | 86 | |
| AK088016.1 | - | 7 - 8 | 1140 | 86 | ||
| AK088729.1 | - | 7 - 8 | 986 | 86 | ||
| AK135521.1 | - | 6 - 7 | 915 | 86 | ||
| AK141253.1 | - | 2 - 3 | 166 | 86 | ||
| AK159689.1 | - | 7 - 8 | 1081 | 86 | ||
| AK167682.1 | - | 7 - 8 | 1137 | 86 | ||
| BC050906.1 | - | 7 - 8 | 1112 | 5721348 | 86 | |
| X15202.1 | - | 6 - 7 | 932 | 86 | ||
| Y00769.1 | - | 7 - 8 | 1027 | 86 | ||
More Information
Set Membership: |
|

Itgb1
Genomic Map