Assay Details
| Gene Symbol: Notum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175263.4 | NP_780472.3 | 9 - 10 | 1089 | 81 | |
| GenBank mRNA | AK019987.1 | - | 9 - 10 | 1089 | 81 | |
| BC025832.1 | - | 4 - 5 | 559 | 5125723 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Notum
Genomic Map