Assay Details
| Gene Symbol: Nploc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195023.1 | NP_001181952.1 | 12 - 13 | 1475 | 69 | |
| NM_199469.2 | NP_955763.1 | 12 - 13 | 1475 | 69 | ||
| GenBank mRNA | AK129375.1 | - | 12 - 13 | 1450 | 69 | |
| AK141998.1 | - | 12 - 13 | 1474 | 69 | ||
| AK149823.1 | - | 12 - 13 | 1447 | 69 | ||
| AK149912.1 | - | 12 - 13 | 1474 | 69 | ||
| BC065156.1 | - | 12 - 13 | 1443 | 5706754 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nploc4
Genomic Map