Assay Details
| Gene Symbol: Nploc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195023.1 | NP_001181952.1 | 15 - 16 | 1753 | 69 | |
| NM_199469.2 | NP_955763.1 | 14 - 15 | 1657 | 69 | ||
| GenBank mRNA | AK129375.1 | - | 15 - 16 | 1728 | 69 | |
| AK149823.1 | - | 15 - 16 | 1725 | 69 | ||
| AK149912.1 | - | 15 - 16 | 1752 | 69 | ||
| AK215926.1 | - | 2 - 3 | 141 | 69 | ||
| BC065156.1 | - | 14 - 15 | 1625 | 5706754 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nploc4
Genomic Map