Assay Details
| Gene Symbol: Nploc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195023.1 | NP_001181952.1 | 9 - 10 | 1102 | 62 | |
| NM_199469.2 | NP_955763.1 | 9 - 10 | 1102 | 62 | ||
| GenBank mRNA | AK129375.1 | - | 9 - 10 | 1077 | 62 | |
| AK141998.1 | - | 9 - 10 | 1101 | 62 | ||
| AK149823.1 | - | 9 - 10 | 1074 | 62 | ||
| AK149912.1 | - | 9 - 10 | 1101 | 62 | ||
| BC065156.1 | - | 9 - 10 | 1070 | 5706754 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nploc4
Genomic Map