Assay Details
| Gene Symbol: Pcyt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024229.2 | NP_077191.2 | 6 - 7 | 573 | 84 | |
| GenBank mRNA | AK140458.1 | - | 6 - 7 | 613 | 84 | |
| AK144544.1 | - | 6 - 7 | 620 | 84 | ||
| BC003473.1 | - | 6 - 7 | 579 | 2650077 | 84 | |
| BC008276.1 | - | 6 - 7 | 573 | 3707732 | 84 | |
More Information
Set Membership: |
|

Pcyt2
Genomic Map